(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:71989-14-5
:
Formula:C₂₃H₂₅NO₆
MDL Number:MFCD00037131
:411.5 g/mol
EINECS:276-251-7
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:148.0 to 152.0 deg-C
- Specific rotation [a]20/D:-22.0 to -27.0 deg (C=1, DMF)
Specification Test Results
Frequently Bought Together