(S)-3-(Benzyloxy)-2-((tert-butoxycarbonyl)amino)propanoic acid ≥98.0% (by titrimetric analysis)
TCIAB1629-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:White to Almost white powder to crystaline
- Purity (HPLC):min. 96.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:57.0 to 62.0 deg-C
- Specific rotation [a]20/D:+21.0 to +24.0 deg (C=2, EtOH)
:
:295.3 g/mol
:245-820-1
:23680-31-1
:C₁₅H₂₁NO₅
:MFCD00066063