Order Entry
Spain
ContactUsLinkComponent
Erioglaucine disodium salt
Erioglaucine disodium salt
Catalog # TCIAB0790-25G
Supplier:  TCI
CAS Number:  
undefined
Erioglaucine disodium salt
Catalog # TCIAB0790-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Red to Dark red to Dark purple powder to crystal
  • Lambda max.:
    627.0 to 631.0 nm (0.02mol/L AcONH4 sol.)
  • Absorbance:
    min. 0.350 (2.5mg/L, 0.02mol/L AcONH4 sol., 627.0 to 631.0 nm) (calcd.on dried substance)
  • Drying loss:
    max. 10.0 %

Specification Test Results

About this item

 :
 :792.84 g/mol
Merck Index:14,01373
EINECS:223-339-8
CAS Number:3844-45-9
Formula:C₃₇H₃₄Na₂N₂O₉S₃
MDL Number:MFCD00012141