Order Entry
Spain
ContactUsLinkComponent
 
Atorvastatin calcium salt trihydrate ≥98.0% (by HPLC, total nitrogen)
 :  TCI
 
Atorvastatin calcium salt trihydrate ≥98.0% (by HPLC, total nitrogen)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 % (calcd.on anh.substance)
  • Specific rotation [a]20/D:
    -7.0 to -10.0 deg (C=1、DMSO) (calcd.on anh.substance)
  • Water:
    3.5 to 5.5 %

 

Frequently Bought Together