Order Entry
Spain
ContactUsLinkComponent
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) ≥90.0% (by HPLC) for biochemical research
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) ≥90.0% (by HPLC) for biochemical research
Catalog # TCIAA2412-5MG
Supplier:  TCI
CAS Number:  
undefined
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) ≥90.0% (by HPLC) for biochemical research
Catalog # TCIAA2412-5MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange powder to crystal
  • Purity (HPLC):
    min. 90.0 area%
  • DNA staining test:
    to pass test

Specification Test Results

About this item

 :
 :350.25 g/mol
Storage Temperature:Freezer
EINECS:249-186-7
CAS Number:28718-90-3
Formula:C₁₆H₁₅N₅·2HCl
MDL Number:MFCD00012681