Triacetin (glycerol triacetate) 98.5-100.5% (by alkalimetry), EMPROVE® ESSENTIAL USP, Ph. Eur., BP suitable for use as excipient, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
Synonyms: Glyceryl triacetate, Propane-1,2,3-triyl triacetate, Glycerol triacetate
CAS Number:102-76-1
:
Formula:C₉H₁₄O₆
MDL Number:MFCD00008716
:218.21 g/mol
Merck Index:13,09664
Boiling Pt:258 °C (1013 hPa)
Melting Pt:–78 °C
Density:1.1596 g/cm³ (20 °C)
Flash Pt:138 °C (closed cup)
Storage Temperature:Ambient
EINECS:203-051-9
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (Alkalimetric, Calculated on Anhydrous Substance):98.5 - 100.5%
- Assay (GC, calc. on anhydrous substance):97.0 - 102.0%
- Identity (IR-spectrum):Passes Test
- Identity (GC):Passes Test
- Appearance:Clear Colorless, Slightly Viscous Liquid, not more Intense in Color than Reference Solution Y₆
- Acidity (Ph. Eur./ USP):passes test
- Refractive Index (n 20/D):1.430 - 1.432
- Density (d 20/20):1.159 - 1.164
- As (Arsenic):≤1.5 ppm
- Acetic Acid (GC):≤0.1%
- Glycerol (GC):≤0.1%
- 1-Monoacetin (GC):≤0.1%
- 2-Monoacetin (GC):≤0.1%
- 1,3-Diacetin (GC):≤0.1%
- 1,2-Diacetin (GC):≤0.1%
- Individual, Unspecified Impurity (GC):≤0.1%
- Sum of all impurities (GC):≤0.2%
- Acetic Acid (Acidity):≤5000 ppm
- Butylacetate (HS-GC):≤5000 ppm
- Other Residual Solvents (ICH Q3C):Excluded by Manufacturing Process
- Water (According to Karl Fischer):≤0.2%
Specification Test Results
Related Information
Frequently Bought Together