
4-(Pentafluorothio)aniline ≥95.0% (by GC, titration analysis)
: TCI
:2993-24-0
:
:C₆H₆F₅NS
:MFCD01076203
:219.18 g/mol
:Refrigerator
:2811
:6.1,III
:GHS06
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 95.0 %
- Purity (Nonaqueous Titration):min. 95.0 %
- Melting point:63.0 to 68.0 deg-C
Frequently Bought Together