Order Entry
Spain
ContactUsLinkComponent
 
 
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate) ≥98.0% (by HPLC)
 :  TCI
 
 
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate) ≥98.0% (by HPLC)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystaline
  • Purity (HPLC):
    min. 98.0 area%

 

Frequently Bought Together