Order Entry
Spain
ContactUsLinkComponent
2-Hydroxy-4-methoxyaniline hydrochloride ≥98.0% (by total nitrogen basis)
Irritant
2-Hydroxy-4-methoxyaniline hydrochloride ≥98.0% (by total nitrogen basis)
Supplier:  TCI
Certificates
undefined
Irritant
2-Hydroxy-4-methoxyaniline hydrochloride ≥98.0% (by total nitrogen basis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light red to Green powder to crystalline
  • Purity (HPLC):
    min. 96.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %

Specification Test Results

Frequently Bought Together