Order Entry
Spain
ContactUsLinkComponent
 
 
Atropine sulphate monohydrate ≥99.0% (by titrimetric analysis)
 :  TCI
 
 
Atropine sulphate monohydrate ≥99.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystaline
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 99.0 %
  • Solubility in Water:
    almost transparency

 

Frequently Bought Together