DL-Alanyl-DL-methionine ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAA0187-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in hot Water:almost transparency
Specification Test Results
About this item
:
:220.29 g/mol
EINECS:217-884-0
CAS Number:1999-43-5
Formula:C₈H₁₆N₂O₃S
MDL Number:MFCD00038166