Order Entry
Spain
ContactUsLinkComponent
Sodium chloride 0.01 mol/l (0.01 N) in aqueous solution
Sodium chloride 0.01 mol/l (0.01 N) in aqueous solution
Catalog # PRLSMC311648.1000
Supplier:  PROLABO CMP
CAS Number:  
Sodium chloride 0.01 mol/l (0.01 N) in aqueous solution
Catalog # PRLSMC311648.1000
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :58.44 g/mol
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477