Order Entry
Spain
ContactUsLinkComponent
VWR®, Silicon standard solution, 10000 mg/L Si in water with hydrofluoric acid (max. 1%) (from (NH4)2SiF6) ARISTAR® standard for ICP
VWR®, Silicon standard solution, 10000 mg/L Si in water with hydrofluoric acid (max. 1%) (from (NH4)2SiF6) ARISTAR® standard for ICP
Catalog # 456012Y
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Silicon standard solution, 10000 mg/L Si in water with hydrofluoric acid (max. 1%) (from (NH4)2SiF6) ARISTAR® standard for ICP
Catalog # 456012Y
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    H₂O/tr HF
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Silicon
  • Symbol:
    Si

Specifications

About this item

(NH₄)₂ SiF₆ in H₂O tr. HF.

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Supplied with certificate of analysis.