Order Entry
Spain
ContactUsLinkComponent
Praseodymium standard solution, 1000 mg/L Pr in dil. nitric acid (from Pr₂O₃) Certipur® standard for ICP, Supelco®
Praseodymium standard solution, 1000 mg/L Pr in dil. nitric acid (from Pr₂O₃) Certipur® standard for ICP, Supelco®
Catalog # 1.70343.0100
Supplier:  Merck
CAS Number:  
undefined
Praseodymium standard solution, 1000 mg/L Pr in dil. nitric acid (from Pr₂O₃) Certipur® standard for ICP, Supelco®
Catalog # 1.70343.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2-3% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ100
  • Element:
    Praseodymium
  • Symbol:
    Pr

Specifications

About this item

Certifications: Traceable to SRM from NIST.