

Fehling's reagent (kit, contains solution I and II) for qualitative determination of reducing sugars
Catalog # 31701.296
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Appearance:Clear blue liquid
- Identification:Passes test
Specification Test Results
About this item
:
Density:1.24 g/cm³ (20 °C)
UN:3266
ADR:8,I
:GHS05
:GHS09