Order Entry
Spain
ContactUsLinkComponent
 
2-Ethylcyclohexanol cis- and trans-mixture ≥97.0% (by GC)
 :  TCI
 
undefined
2-Ethylcyclohexanol cis- and trans-mixture ≥97.0% (by GC)
 :  TCI

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 % (total of isomers)

 

Frequently Bought Together