Order Entry
Spain
ContactUsLinkComponent
Ruthenium ≥99.9% (metals basis), cubes, Length: 6 mm (0.24 in)
Ruthenium ≥99.9% (metals basis), cubes, Length: 6 mm (0.24 in)
Catalog # 45484.KF
CAS Number:  
Ruthenium ≥99.9% (metals basis), cubes, Length: 6 mm (0.24 in)
Catalog # 45484.KF
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

CAS Number:7440-18-8
 :
Formula:Ru
MDL Number:MFCD00011207
 :101.07 g/mol
Storage Temperature:Ambient
EINECS:231-127-1