Order Entry
Spain
ContactUsLinkComponent
 
Potassium sulphate, tablets, PS KJELTABS mineralisation catalyst
Catalog # THOMAA23
Supplier:  THOMPSON & CAPPER
CAS Number:  
Potassium sulphate, tablets, PS KJELTABS mineralisation catalyst
Catalog # THOMAA23
Supplier:  THOMPSON & CAPPER
CAS Number:  

Specifications

  • Pk:
    1.000 Tab.
  • Seq:
    0980

Specifications

About this item

 :
 :174.26 g/mol
Merck Index:13,07759
Boiling Pt:1698 °C (1013 hPa)
Melting Pt:1069 °C
Density:2.66 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-915-5
CAS Number:7778-80-5
Formula:K₂SO₄
MDL Number:MFCD00011388