Order Entry
Spain
ContactUsLinkComponent
Phenol liquefied/tris-Cl, Gen-Apex for DNA extraction
Toxic
Corrosive
Health
Phenol liquefied/tris-Cl, Gen-Apex for DNA extraction
Catalog # 33595.188
Supplier:  VWR Chemicals
CAS Number:  
undefined
Toxic
Corrosive
Health
Phenol liquefied/tris-Cl, Gen-Apex for DNA extraction
Catalog # 33595.188
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
Storage Temperature:Ambient
EINECS:203-632-7
UN:2821
ADR:6.1,II
 :GHS06
 :GHS05
 :GHS08
CAS Number:108-95-2
Formula:C₆H₅OH