Order Entry
Spain
ContactUsLinkComponent
Dow Corning® 1107 water repellent and powder treatment
Dow Corning® 1107 water repellent and powder treatment
Catalog # 632198T
Supplier:  DOW CORNING
CAS Number:  
Dow Corning® 1107 water repellent and powder treatment
Catalog # 632198T
Supplier:  DOW CORNING
CAS Number:  

Specifications

  • Pack type:
    Plastic container
  • Pk:
    200 kg
  • Seq:
    4676

Specifications

About this item

 :
Boiling Pt:>140 °C (0.003 hPa)
Melting Pt:–55 °C
Density:0.971 g/cm³ (25 °C)
Flash Pt:316 °C (closed cup)
Storage Temperature:Ambient
CAS Number:63148-57-2