About this item
The Nalgene™ multi-tiered portfolio provides bioproduction facilities with a variety of high quality, sterile storage and transport solutions. Choose the appropriate product necessary to suit any application, through the step-down process and into the cleanroom.
- Eliminates costly washing, depyrogenation and autoclaving steps
- Made from durable, break-resistant polyethylene terephthalate copolyester, glycol modified (PETG)
- Suitable for frozen storage down to –40 °C
- Heat-shrink band around closure and neck for tamper-evident seal
- Leakproof bottles and HDPE closures provided
- Sterile and non pyrogenic
- Supported by extensive validation documentation
Bottles are washed with USP water for injection and handled post-washing in class 5 cleanrooms with class 4 packaging/inspection hoods to produce these very low particulate products. Certified and lot-to-lot tested to contain less than ¹/₃ the particulates allowable according to USP <788> to limit potential particulate contamination. Sterile, ready to use products with a sterility assurance level of 10⁻⁶ inside the container/closure, including two heat-sealed sterile barriers to protect the product prior to use.
Products are ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates. They are also well suited for the preparation and containment of buffers, cultures or prolonged storage of pH-sensitive liquids such as culture media.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- 216-2295
- 216-2294
- 216-2292
- 216-2290
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement