About this item
Fibroblast growth factor 7 (FGF-7), also known as keratinocyte growth factor (KGF), is a potent mitogen that regulates epithelial cell migration and differentiation. FGF-7 is produced by mesenchymal cells and binds in high affinity to a splice variant of FGF receptor 2 (FGFR2-IIIb). The mitogenic activity of FGF-7 acts predominantly on keratinocytes, but not on fibroblast or endothelial cells. FGF-7 expression is upregulated after acute and chronic injury, suggesting that FGF-7 functions during the healing of injured epithelial cells. FGF-7 also induces the formation of the apical ectodermal ridge during limb development.
- High quality
- Low endotoxin
- Affordable
FGF7 expression is upregulated after acute and chronic injury, suggesting that FGF7 functions during the healing of injured epithelial cells. FGF7 also induces the formation of the apical ectoderm ridge during limb development.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity Assay: 4MBr-5 cell proliferation, ED₅₀ ≤60 ng/ml, Bioactivity ≥1,7×10⁴ units/mg
- Gene ID:P21781
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:KGF
- Protein/peptide name:FGF-7 / KGF (AF)
- Purity:≥90%
- Molecular weight:19 kDa
- Sequence:MACNDMTPEQM ATNVNCSSPE RHTRSYDYME GGDIRVRRLF CRTQWYLRID KRGKVKGTQE MKNNYNIMEI RTVAVGIVAI KGVESEFYLA MNKEGKLYAK KECNEDCNFK ELILENHYNT YASAKWTHNG GEMFVALNQK GIPVRGKKTK KEQKTAHFLP MAIT
- Endotoxin level:Low
- Formulation:20 mM Sodium phosphate, 0,5 M Sodium chloride, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay