Mouse Recombinant IL-27/P28 (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
The p28 subunit of interleukin 27 (IL-27), also known as interleukin 30 (IL-30), is a member of the interleukin 12 (IL-12) family of cytokines. p28 is a secreted polypeptide that associates with the Epstein-Barr virus induced gene 3 (EBI3) to form the IL-27 cytokine heterodimer complex. IL-27 functions as a proinflammatory cytokine that induces immunomodulatory effects in naïve CD4+ T cells, mast cells, and monocytes. p28 can also form a complex with cytokine-like factor 1 (CLF), that is secreted by dendritic cells, to regulate natural killer (NK) and T cell functions.
- High quality
- Low endotoxin
- Affordable
Caution: This product is for research use only and is not for use in humans.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:Q8K3I6
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:P28|IL-27 p28 subunit|IL-30
- Protein/peptide name:IL-27/P28
- Purity:≥95%
- Molecular weight:23,7 kDa
- Sequence:MFPTDPLSLQ ELRREFTVSL YLARKLLSEV QGYVHSFAES RLPGVNLDLL PLGYHLPNVS LTFQAWHHLS DSERLCFLAT TLRPFPAMLG GLGTQGTWTS SEREQLWAMR LDLRDLHRHL RFQVLAAGFK CSKEEEDKEE EEEEEEEEKK LPLGALGGPN QVSSQVSWPQ LLYTYQLLHS LELVLSRAVR DLLLLSLPRR PGSAWDS
- Endotoxin level:Low
- Formulation:10 mM Sodium bicarbonate, pH 8,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Related Information
Literature reference - PubMed ID 21057510 Literature reference - PubMed ID 24021664
Additional Product Details
Frequently Bought Together