Order Entry
Spain
ContactUsLinkComponent
Mouse Recombinant Leptin (from E. coli)
Mouse Recombinant Leptin (from E. coli)
Catalog # SHBT200-26AF-200UG
CAS Number:  
undefined
Mouse Recombinant Leptin (from E. coli)
Catalog # SHBT200-26AF-200UG
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    200 µG
  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Seq:
    1358
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤1
  • Biological activity:
    Bioactivity: No biological activity data is available at this time
  • Gene ID:
    Q544U0
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    Obesity protein
  • Protein/peptide name:
    LEPTIN (AF)
  • Purity:
    ≥95%
  • Molecular weight:
    16,1 kDa
  • Sequence:
    MVPIQKVQDD TKTLIKTIVT RINDISHTQS VSAKQRVTGL DFIPGLHPIL SLSKMDQTLA VYQQVLTSLP SQNVLQIAND LENLRDLLHL LAFSKSCSLP QTSGLQKPES LDGVLEASLY STEVVALSRL QGSLQDILQQ LDVSPEC
  • Endotoxin level:
    Low
  • Formulation:
    10 mM Sodium phosphate, pH 7,5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

Specifications

About this item

Leptin is a hormone that is produced by adipose tissue and plays critical roles in the physiologic regulation of body weight. Leptin acts through the leptin receptor (LEPR) to regulate adipose mass by inhibiting hunger and balancing energy usage. Leptin mutations cause severe hereditary obesity and hypogonadism in rodents and humans. Leptin also has thermogenic actions, regulates enzymes of fatty acid oxidation, and is involved in hematopoiesis, angiogenesis, wound healing, inflammation, and immune responses.

  • Leptin is a hormone that is produced by adipose tissue and plays critical roles in the physiologic regulation of body weight. Leptin acts through the leptin receptor (LEPR) to regulate adipose mass by inhibiting hunger and balancing energy usage. Leptin mutations cause severe hereditary obesity and hypogonadism in rodents and humans. Leptin also has thermogenic actions, regulates enzymes of fatty acid oxidation, and is involved in hematopoiesis, angiogenesis, wound healing, inflammation, and immune responses.
Caution: This product is for research use only and is not for use in humans.