Human Recombinant GACRP-30 (from E. coli)
The globular subunit of adipocyte complement-related protein of 30 kDa (ACRP-30) is a naturally occurring cleavage product of adiponectin, a protein made exclusively by adipocytes. ACRP-30 is an abundant serum protein and plays an important role in hyperglycemia, insulin resistance, and fatty acid oxidation. ACRP-30 signals through adiponectin receptor 1 (AdipoR1) and adiponectin receptor 2 (AdipoR2).
- High quality
- Low endotoxin
- Affordable
: This product is for research use only and is not for use in humans.
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity Assay: Inhibition of M1 cell proliferation, ED₅₀ ≤2000 ng/ml, Bioactivity ≥500 units/mg
- Gene ID:Q15848
- Reconstitution instructions:Sterile 10 mM sodium phosphate, 0,5 mM DTT, pH 7,5 at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:adipolean|Adiponectin|apm-1
- Protein/peptide name:GACRP-30 (AF)
- Purity:≥90%
- Molecular weight:16,7 kDa
- Sequence:MKGEPGEGAY VYRSAFSVGL ETYVTIPNMP IRFTKIFYNQ QNHYDGSTGK FHCNIPGLYY FAYHITVYMK DVKVSLFKKD KAMLFTYDQY QENNVDQASG SVLLHLEVGD QVWLQVYGEG ERNGLYADND NDSTFTGFLL YHDTN
- Endotoxin level:Low
- Formulation:10 mM Sodium phosphate, 0,5 mM DTT, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Frequently Bought Together