Industrial masking tapes, 401E
: 3M
401E masking tape is a smooth crepe paper tape with rubber adhesive. It offers high tensile strength, with a thickness of 0,170 mm.
- Ideal in painting baking processes
- May be exposed to temperatures up to 140 °C
- Will not tear
- Resistant to solvents and moisture, high temperatures
- Designed for indoor use
- Tensile strength: 100 [Newton/25 mm]
- Crepe paper back material
Suitable for all oven-drying systems, infrared, or heat lamps.
Frequently Bought Together