

Dichloromethane ≥99.5% (by GC) inhibitor-free, Selectophore™ ionophore, Supelco®
Catalog # 91301-500ML
Supplier: Merck
CAS Number:
Specifications
- Pk:500 mL
- Seq:2725
Specifications
About this item
:
:84.93 g/mol
Melting Pt:−97 °C
UN:1593
ADR:6.1,III
:GHS07
:GHS08
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881