Anthranilamide ≥99.0% (by HPLC) matrix substance for MALDI-MS, Supelco®
76884-1G
: Merck
:
- Pk:1 g
- Seq:3047
- Appearance (Color):White
- Appearance (Form):Powder or Crystals or Flakes
- Titration (NT) HCLO4 0.1M:99 - 101 %
- Purity (HPLC area %):≥ 99.0 %
- Proton NMR Spectrum:Conforms to structure
- Metal trace Analysis (ICP):Corresponds to requirements
- BARIUM (ICP):≤ 5.0 mg/kg
- CALCIUM (ICP):≤ 5.0 mg/kg
- CADMIUM (ICP):≤ 5.0 mg/kg
- COBALT (ICP):≤ 5.0 mg/kg
- CHROMIUM (ICP):≤ 5.0 mg/kg
- COPPER (ICP):≤ 5.0 mg/kg
- IRON (ICP):≤ 5.0 mg/kg
- POTASSIUM (ICP):≤ 50 mg/kg
- MAGNESIUM (ICP):≤ 5.0 mg/kg
- MANGANESE (ICP):≤ 5.0 mg/kg
- SODIUM (ICP):≤ 150 mg/kg
- NICKEL (ICP):≤ 5.0 mg/kg
- LEAD (ICP):≤ 5.0 mg/kg
- ZINC (ICP):≤ 5.0 mg/kg
- RECOMMENDED RETEST PERIOD:48 MONTHS
:
:136.15 g/mol
:88-68-6
:C₇H₈N₂O
:MFCD00007981