Avantor® Hichrom, Chromatography Tubing, Stainless Steel
Supplier: Avantor
Certificates
About this item
Tubings are made of 316 stainless steel.
- Resistant to chemicals
The precision bore is ¹/₁₆" type tubing.
Frequently Bought Together
Tubings are made of 316 stainless steel.
The precision bore is ¹/₁₆" type tubing.