Order Entry
Spain
ContactUsLinkComponent
10-Hydroxynaltrexone solution, certified reference material, Supelco®
10-Hydroxynaltrexone solution, certified reference material, Supelco®
Catalog # IMPN-007-02-1M
Supplier:  Merck
CAS Number:  
10-Hydroxynaltrexone solution, certified reference material, Supelco®
Catalog # IMPN-007-02-1M
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    100 mug/mL in methanol
  • Pk:
    1 mL
  • Standards:
    certified reference material
  • CAS No.:
    60767-82-0
  • Description:
    10-Hydroxynaltrexone solution
  • Description:
    10-Hydroxynaltrexone solution
  • Pk Info:
    ampoule of 1 ml

Specifications

About this item