Paraxanthine solution, certified reference material, Supelco®
IMPC-051-03-1M
: Merck
:
- Concentration:1.0 mg/mL in methanol
- Pk:1 mL
- Standards:certified reference material
- CAS No.:611-59-6
- Description:Paraxanthine solution
- Description:Paraxanthine solution
- Pk Info:ampoule of 1 ml