Order Entry
Spain
ContactUsLinkComponent
Supelco® Organic Reference Standard, Urea
Supelco® Organic Reference Standard, Urea
Catalog # 56180-1G
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Urea
Catalog # 56180-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 g
  • Standards:
    Analytical standard
  • CAS No.:
    57-13-6
  • Description:
    Urea
  • Storage conditions:
    RT
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00008022
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Carbamide; Carbonyldiamide
  • Melting point:
    132 - 135 °C (lit.)
  • Formula:
    NH₂CONH₂
  • Molecular weight:
    60,06
  • Density:
    1,335 g/ml at 25 °C (lit.)

Specifications

About this item