Biocytin hydrazide
Catalog # 90060.
Supplier: Biotium
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
- Environmentally Preferable:
Specifications
About this item
:
:386.515 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:102743-85-1
Formula:C₁₆H₃₀N₆O₃S