Supelco® Organic Reference Standard, Xanthotoxin
Xanthotoxin, Analytical standard, 50 mg
Catalog # 56448-50MG
Supplier: Merck
Jump to:
Product Details & Documents
- GHS signal: GHS07
Caution: P202 - P261 - P280 - P301 + P312 - P302 + P352 - P308 + P313
Specifications
Specifications
- Pk:50 mg
- Standards:Analytical standard
- Description:Xanthotoxin
- Cat. No.:56448-50MG
- M-Clarity quality level:MQ100
- Storage conditions:2…8 °C
- Hazard class description:H302 - H317 - H341 - H351
- MDL No.:MFCD00005009
- Melting point:148 - 150 °C (lit.)
- Synonym(s):8-Methoxypsoralen; 8-MOP; 9-Methoxyfuro[3,2-g][1]benzopyran-7-one; Ammoidin; Methoxsalen; Xanthotoxin
- Molecular weight:216,19
- CAS No.:298-81-7
- Shipping conditions:STD



