Supelco® Organic Reference Standard, Pyrogallol
Catalog # 06931-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:Analytical standard
- CAS No.:87-66-1
- Description:Pyrogallol
- Environmentally Preferable:
- Storage conditions:2...8 °C
- Hazard class description:H302 + H312 + H332 - H341 - H412
- Shipping conditions:STD
- MDL No.:MFCD00002192
- M-Clarity quality level:MQ100
- Synonym(s):1,2,3-Trihydroxybenzene; 2,3-Dihydroxyphenol; Pyrogallic acid
- Melting point:132...134 °C; 133...134 °C (lit.)
- Formula:C₆H₃(OH)₃
- Molecular weight:126,11
- Storage temperature:2...8 °C
- Flash point:Not applicable
- Boiling point:309 °C (lit.)
Specifications
About this item
- GHS signal: GHS08, GHS07
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.
Caution: P273 - P280 - P301 + P312 - P302 + P352 + P312 - P304 + P340 + P312 - P308 + P313