Order Entry
Spain
Orders LinkContactUsLinkComponent
Fructose assay kit, Supelco®

Fructose assay kit, Supelco®

Fructose Assay Kit, Sufficient for 20 assays, 1 KIT
Catalog # FA20-1KT
Supplier:   Merck
Fructose assay kit, Supelco®
Catalog # FA20-1KT
Supplier:  Merck
CAS Number:  
Fructose assay kit, Supelco®

Fructose Assay Kit, Sufficient for 20 assays, 1 KIT

CAS Number:  
Price
301,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

This kit is for the quantitative, enzymatic determination of fructose in food and other materials.

Fructose is phosphorylated by adenosine triphosphate (ATP) in the reaction catalyzed by hexokinase. Fructose 6-phosphate is converted to glucose 6-phosphate by phosphoglucose isomerase (PGI). Glucose-6-phosphate (G6P) is then oxidized to 6-phosphogluconate in the presence of nicotinamide adenine dinucleotide (NAD) in the reaction catalysed by glucose-6-phosphate dehydrogenase (G6PDH). During this oxidation, an equimolar amount of NAD is reduced to NADH. The consequent increase in absorbance at 340 nm is directly proportional to fructose concentration.

Kit components

Fructose standard 500 mg
Glucose assay reagent 1 ml/vial
Phosphoglucose isomerase 1 ml/vial

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    Fructose Assay Kit
  • Pk Info:
    Sufficient for 20 assays
  • Cat. No.:
    FA20-1KT
  • M-Clarity quality level:
    MQ100

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.