Organic reference standard, disperse yellow 3, analytical standard, Supelco®
Catalog # 11344-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:2832-40-8
- Description:Disperse Yellow 3, ≥95,0% (HPLC)
- M-Clarity quality level:MQ100
- Melting point:268...270 °C (lit.)
- Formula:C₁₅H₁₅N₃O₂
- Description:Disperse Yellow 3
- Form:Neat
Specifications
About this item
Disperse Yellow 3 is an yellow azo dye which is majorly used in textile industry. It also has carcinogenic properties.
It is used to understand that Phanerochaete chrysosporium can mineralise azo dyes with radiolabelled substrate.
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.