Supelco® Organic Reference Standard, Betulin
Catalog # 92648-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:Analytical standard
- CAS No.:473-98-3
- Description:Betulin
- Storage conditions:RT
- Hazard class description:H371
- Shipping conditions:STD
- MDL No.:MFCD00016802
- M-Clarity quality level:MQ100
- Synonym(s):Betulinic alcohol; Betulinol; Betulol; Lup-20(29)-ene-3β,28-diol; Trochol
- Melting point:256 - 257 °C (lit.)
- Molecular weight:442,72
Specifications
About this item
- GHS signal: GHS08
Caution: P260 - P264 - P270 - P308 + P311 - P405 - P501