Order Entry
Spain
ContactUsLinkComponent
Supelco® Organic Reference Standard, Avenanthramide B
Supelco® Organic Reference Standard, Avenanthramide B
Catalog # 93105-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Avenanthramide B
Catalog # 93105-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • Description:
    Avenanthramide B
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H302
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD23380832
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    5-Hydroxy-2-{[(2E)-3-(4-hydroxy-3-methoxyphenyl)-1-oxo-2-propenyl]amino}benzoic acid; N-[4′-Hydroxy-3′-methoxy-(E)-cinnamoyl]-5-hydroxyanthranilic acid; Avenanthramide Bf
  • Molecular weight:
    329,30

Specifications

About this item

  • GHS signal: GHS07