Order Entry
Spain
ContactUsLinkComponent
Supelco® Organic Reference Standard, α-Tocotrienol
Supelco® Organic Reference Standard, α-Tocotrienol
Catalog # 07205-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, α-Tocotrienol
Catalog # 07205-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    58864-81-6
  • Description:
    α-Tocotrienol
  • Storage conditions:
    –20 °C
  • Shipping conditions:
    DICE
  • MDL No.:
    MFCD11045308
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    ᴅ-α-Tocotrienol; (R)-α-Tocotrienol; [R-(E,E)]-3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyl-3,7,11-tridecatrienyl)-2H-1-benzopyran-6-ol
  • Formula:
    C₂₉H₄₄O₂
  • Molecular weight:
    424,66
  • Storage temperature:
    –20 °C
  • Flash point:
    Not applicable

Specifications

About this item