Supelco® Organic Reference Standard, Agnuside
12274-10MG
: Merck
:
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:11027-63-7
- Description:Agnuside
- Environmentally Preferable:
- Storage conditions:2...30 °C
- Hazard class description:H302
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Synonym(s):(1S,4aR,5S,7aS)-1,4a,5,7a-Tetrahydro-5-hydroxy-7-[[(4-hydroxybenzoyl)oxy]methyl]cyclopenta[c]pyran-1-yl β-ᴅ-glucopyranoside; Buddlejoside A
- Formula:C₂₂H₂₆O₁₁
- Molecular weight:466,44
- Flash point:Not applicable
- Form:Neat
- GHS signal: GHS07
: P301 + P312 + P330