Organic reference standard, 6-Methyl-5-hepten-2-one, analytical standard, Supelco®
Catalog # 67397-1ML
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:110-93-0
- Description:6-Methyl-5-hepten-2-one, ≥97,0% (GC)
- Formula:C₈H₁₄O
- Description:6-Methyl-5-hepten-2-one
- Flash point:56 °C
- Boiling point:73 °C/18 mm Hg (lit.)
- Form:Neat