Supelco® Organic Reference Standard, 1-Deoxynojirimycin
Catalog # 08012-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:19130-96-2
- Description:1-Deoxynojirimycin
- Environmentally Preferable:
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD00063474
- M-Clarity quality level:MQ100
- Synonym(s):1,5-Dideoxy-1,5-imino-ᴅ-sorbitol
- Formula:C₆H₁₃NO₄
- Molecular weight:163,17
- Flash point:Not applicable