Order Entry
Spain
ContactUsLinkComponent
Lucifer Yellow Cadaverine fluorescent dye, for histology
Lucifer Yellow Cadaverine fluorescent dye, for histology
Catalog # 80018.
Supplier:  Biotium
CAS Number:  
undefined
Lucifer Yellow Cadaverine fluorescent dye, for histology
Catalog # 80018.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :533.66 g/mol
Storage Temperature:Refrigerator
CAS Number:149733-79-9
Formula:C₁₇H₁₇K₂N₃O₈S₂