Order Entry
Spain
ContactUsLinkComponent
Octadecyl rhodamine B chloride (R 18) membrane stain, for histology
Irritant
Octadecyl rhodamine B chloride (R 18) membrane stain, for histology
Catalog # 60033.
Supplier:  Biotium
CAS Number:  
undefined
Irritant
Octadecyl rhodamine B chloride (R 18) membrane stain, for histology
Catalog # 60033.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :731.50 g/mol
 :GHS07
Storage Temperature:Freezer
CAS Number:65603-19-2
Formula:C₄₆H₆₇ClN₂O₃
MDL Number:MFCD00467892