Order Entry
Spain
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, P-I-A-N-O Kit
Catalog # 44594-U
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, P-I-A-N-O Kit
Catalog # 44594-U
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Standards:
    Analytical standard
  • Description:
    P-I-A-N-O Kit
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H225 - H302 - H304 - H315 - H317 - H318 - H335 - H336 - H340 - H350 - H361fd - H373 - H373 - H410
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100

Specifications

About this item

  • GHS signal: GHS08, GHS07, GHS09
Caution: P210 - P273 - P280 - P301 + P310 - P305 + P351 + P338 - P331