Organic analytical standards, Methanol, reference material, Honeywell Fluka™
HONCR1587-5ML
:
- Pack type:Glass Bottle
- Pk:5 mL
- Standards:Analytical Standard
- CAS No.:67-56-1
- Description:Methanol
- Formula:CH₃OH
- Storage temperature:Ambient
- Description:Methanol
- Form:Liquid
High quality organic reference materials produced according to ISO 9001.
: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.