Order Entry
Spain
ContactUsLinkComponent
Pesticide reference standard, Toxaphene solution, certified reference material, Supelco®
Pesticide reference standard, Toxaphene solution, certified reference material, Supelco®
Catalog # 48103.
Supplier:  Merck
CAS Number:  
Pesticide reference standard, Toxaphene solution, certified reference material, Supelco®
Catalog # 48103.
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml in isooctane
  • Pk:
    1
  • Standards:
    Certified reference material
  • Description:
    Toxaphene solution
  • Storage temperature:
    RT
  • Description:
    Toxaphene solution
  • Pk Info:
    1 ml/ampule

Specifications

About this item

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.