Supelco® PESTANAL® Organic Reference Standard, Pyrazoxyfen
Catalog # 32437-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:71561-11-0
- Description:Pyrazoxyfen
- Storage conditions:2...30 °C
- Hazard class description:H302 - H410
- Shipping conditions:STD
- MDL No.:MFCD00274563
- M-Clarity quality level:MQ100
- Synonym(s):2-[4-(2,4-Dichlorobenzoyl)-1,3-dimethylpyrazol-5-yloxy]acetophenone
- Formula:C₂₀H₁₆Cl₂N₂O₃
- Molecular weight:403,26
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS07, GHS09
Caution: P273 - P301 + P312 + P330