Order Entry
Spain
ContactUsLinkComponent
Organic reference standard, Hexachlorobenzene solution, certified reference material, Supelco®
Organic reference standard, Hexachlorobenzene solution, certified reference material, Supelco®
Catalog # SUPL40008
Supplier:  Merck
CAS Number:  
Organic reference standard, Hexachlorobenzene solution, certified reference material, Supelco®
Catalog # SUPL40008
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml in acetone
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • CAS No.:
    118-74-1
  • Description:
    Hexachlorobenzene solution
  • Formula:
    C₆Cl₆
  • Storage temperature:
    2...8 °C
  • Description:
    Hexachlorobenzene solution
  • Pk Info:
    1 ml/ampule

Specifications

About this item